Learn More
Pyramex™ Venture 3™ Clear Anti-Fog Lens with Black Temples
Soft dual-injected rubber temple tips combined with fully adjustable straight back temples reinforce high comfort and fit
Supplier: Pyramex™ SB5720DT
Description
- Soft and adjustable ventilated nose piece
- Top of frame ventilated allowing air to circulate
- 9.5 base curve lens provides excellent side protection
- Scratch-resistant polycarbonate lens provide 99% UVA/B/C protection
Specifications
| Anti-fog, Scratch-resistant | |
| Black | |
| Polycarbonate |
| Gray | |
| Fully adjustable straight back temples |