Learn More
Pyramex™ V2G™ Safety Eyewear
Catalog No. 191303501
Change view
Click to view available options
Lens Tint:
Clear
Gray
Mirror, Indoor/Outdoor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
191303501 Clear
191303502 Gray
191303503 Mirror, Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191303501 Supplier Pyramex™ Supplier No. GB1810ST
Only null left
Foam padding forms a perfect seal to keep out dust and airborne particles.
  • Lenses are treated to prevent fogging and self seals small scratches
  • 9.75 base curved lens provides full side protection
  • Headband features quick fit retainer
  • Lenses provide 99% protection against harmful UV rays
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Goggles
Includes Black strap; Foam padding; Interchangeable temples
Material Polycarbonate
UV Protection UV400