Learn More
Pyramex™ Venture II Readers Safety Glasses
Venture II Readers Safety Glasses with base curved lens provide full side protection without distracting seams or traditional type side shields.
Supplier:  Pyramex™ SB1810R25
 View more versions of this product
Catalog No. 19-121-924
Only null left
Explore available promotions
Description
Description
  • Nose buds secure the glass and prevent slipping while providing a pillow soft fit
  • Fully adjustable temples - 4 size settings
  • Lenses provide 99% protection against harmful UV rays
  • Lenses are coated for superior scratch resistance
  • Strategic placement of lens insert is out of user's direct line of vision
Specifications
Specifications
Scratch-resistant, UV-treated, Impact-resistant
Clear, Gray, Amber, Indoor/Outdoor Mirror
For indoor applications
+2.50
Polycarbonate
Half Frame
Magnifying
Reader Spectacles
Black
Reusable
U6
Product Suggestions
Product Suggestions