Learn More
Pyramex™ SX3 Plus™ Safety Glasses
Catalog No. p-9042006
Change view
Click to view available options
Lens Tint:
Clear
Gray
Catalog No. Lens Tint
296359664 Gray
296359663 Clear
2 options
Catalog No. 296359664
Supplier: Pyramex™ SB4620STP
Only null left
Sports-look safety spectacle with superior eye protection.
  • Open temples allow clip on cord option for goggle-like fit
  • Unique slim temple design with co-injection temple tips
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection.
  • Soft foam padding provides comfort fit; elastic strap
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) Eye protection
Frame Color Black
Temple Style Adjustable
Lens Material Polycarbonate