missing translation for 'onlineSavingsMsg'
Learn More

Safety Choice™ PVC Dipped Gloves with Interlock Liner

Catalog No. 19013546
Change view
Click to view available options
Exterior Finish:
No Texture
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Exterior Finish
19013546 No Texture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19013546 Supplier Safety Choice™ Supplier No. 588015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single dipped gloves feature an interlock lining that dries quickly, sheds minimal lint, and increases user comfort. Available in either a smooth or rough finish for enhanced grip capabilities.

  • Single dipped PVC gloves with interlock liner
  • Enhanced grip capabilities when handling wet or oily materials
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Straight cuff protects the forearm from liquid run-off
  • Black color helps to hide dirt and stains

Specifications

Coating Material PVC
Color Black, Natural
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material PVC
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Construction, Mining, Refining, and Petrochemicals
Glove Cuff Style Knit
Liner Material Cotton
Show More Show Less