missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PV-1 Performance Verification Solution

Catalog No. 13400500
Change view
Click to view available options
Type:
Performance Verification Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13400500 Performance Verification Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13400500 Supplier Thermo Scientific™ Supplier No. CHEMPV18KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ PV-1 Performance Verification Solution for verifying the accuracy of pedestal pathlengths

  • PV-1 solution is used to verify the reproducibility and accuracy of Thermo Scientific™ NanoDrop™ One/OneC pedestal pathlengths
  • Solution has potassium nitrate and nicotinic asid components
  • Once an ampoule is opened, the solution should be used within 60 minutes
  • Each pack comes with eight PV-1 Performance Verification ampoules

Specifications

Type Performance Verification Solution
For Use With (Equipment) Thermo Scientific™ NanoDrop™ One/OneC
Quantity 8 Ampoules/ Pack
Storage Requirements Store at room temperature