Learn More
PURUS™ Frames for EnviroTack™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
Supplier:  PURUS™ PF 2847 01 W 00
 View more versions of this product
Catalog No. 19125103
Only null left
Description
Description
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS™ EnviroTack™ mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications
Chemical Identifiers
White
Specifications
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
711 x 1194 mm
660 x 1143 mm PURUS™ Biohybrid™ Mats
Polystyrene
3.175 mm
White
Product Suggestions
Product Suggestions