missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
GREENER_CHOICE

Catalog No. 19126394
Change view
Click to view available options
Color:
Blue
Clear
Gray
White
Dimensions (L x W):
18 x 36 in.
18 x 45 in.
24 x 30 in.
24 x 36 in.
24 x 60 in.
26 x 45 in.
36 x 36 in.
36 x 45 in.
36 x 60 in.
Surface Coating:
High Tack
Regular Tack
Quantity:
30 Sheets/Mat, 4 Mats/Cs.
30 Sheets/Mat, 8 Mats/Cs.
38 product options available for selection
Product selection table with 38 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W) Surface Coating Quantity
19126394 White 18 x 45 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125814 Gray 18 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125826 Blue 26 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125832 Blue 36 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125808 Gray 18 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125806 White 18 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125812 White 18 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125807 Blue 18 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125813 Blue 18 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125800 White 24 x 30 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125801 Blue 24 x 30 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125819 Blue 24 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125818 White 24 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125827 Gray 26 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125841 Blue 36 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125842 Gray 36 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125833 Gray 36 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125838 Blue 36 x 60 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125840 White 36 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125839 Gray 36 x 60 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125837 White 36 x 60 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125860 White 18 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125831 White 36 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125862 Blue 36 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125861 Blue 18 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125825 White 26 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
17125730 Gray 36 x 45 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125820 Gray 24 x 36 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125851 Clear 36 x 45 in. Regular Tack 30 Sheets/Mat, 8 Mats/Cs.
19125798 Gray 18 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125799 Gray 36 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125850 Blue 24 x 60 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125950 Blue 26 x 45 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19126392 White 36 x 45 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19126393 Blue 18 x 45 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125924 Blue 36 x 45 in. High Tack 30 Sheets/Mat, 8 Mats/Cs.
19125802 Gray 24 x 30 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
19125965 Gray 24 x 36 in. Regular Tack 30 Sheets/Mat, 4 Mats/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
38 options
Catalog No. 19126394 Supplier PURUS™ Supplier No. PM 1845 34 W

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels

  • Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers

Certifications

Manufactured in ISO 9001 certified facility

Specifications

Quantity 30 Sheets/Mat, 4 Mats/Cs.
Surface Coating Regular Tack
Type Contamination Control Mat
Dimensions (L x W) 18 x 45 in.
Color White
Thickness (English) 1.4 mil
Material LDPE Blend
For Use With (Application) For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications