Learn More
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
GREENER_CHOICE
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
Supplier:  PURUS™ PM 2436 34 G
 View more versions of this product
Catalog No. 19125965
Only null left
Description
Description
  • Earth Friendly containing GMO free, renewable plant based Cardia Biohybrid&trade material that reduces CO2 emissions
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Certifications
Manufactured in ISO 9001 certified facility
Specifications
Chemical Identifiers
LDPE Blend
Contamination Control Mat
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
Specifications
30 Sheets/Mat, 4 Mats/Cs.
Regular Tack
Single-use
Gray
LDPE Blend
Contamination Control Mat
24 x 36 in.
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
1.4 mil
Product Suggestions
Product Suggestions