Learn More
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
GREENER_CHOICE
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels
Supplier:  PURUS™ PS 3636 38 W
 View more versions of this product
Catalog No. 19125840
Only null left
Description
Description
  • Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Certifications
Manufactured in ISO 9001 certified facility
Specifications
Chemical Identifiers
LDPE Blend
Contamination Control Mat
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
Specifications
36 x 36 in.
30 Sheets/Mat, 8 Mats/Cs.
LDPE Blend
Film: 1.3 ± 0.2 mil, Adhesive: 0.3 mil
Film: 33 μm Adhesive: 8 μm
Contamination Control Mat
Regular Tack
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
White
Manufactured in an ISO 9001 certified facility
Product Suggestions
Product Suggestions