Learn More
Puritan™ HydraFlock™ Flocked Swabs
For collection and release of biological specimens
 View more versions of this product
Catalog No. 22025199
Only null left
Description
Description
A new era in flock swab technology. HydraFlock provides superior collection and release of specimens. The unique microstructure of Hydra’s multi-length fibers are designed to enhance rapid absorption and quick release of biological specimens.; For buccal cell collection; Specimen collection; Tip material is comprised of polyester bicomponent fiber; Made in USA
Specifications
Chemical Identifiers
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Sterile
Flocked
Swab
Polystyrene
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2 cm
15.2 cm
1.6 cm
0.14 in.
Polystyrene
Flocked
0.1 in.
For Nasopharyngeal and Urethral cell collection, rapid diagnostic testing, specimen collection, forensic evidence collection
0.254 cm
5.99 in.
0.66 in.
0.35 cm
Sterile
Ultrafine
Product Suggestions
Product Suggestions