Learn More
Puritan™ ESK™ Sampling Kits with Pre-filled Neutralizing Buffer
For environmental surface sampling with Pre-filled Neutralizing Buffer
Supplier:  Puritan™ 25-83004PDBNB
 View more versions of this product
Catalog No. 14-960-222
Only null left
Explore available promotions
Description
Description
For environmental surface sampling; Intended for the detection of microorganisms found on dairy and food equipment disinfected with chlorine or quaternary ammonium compounds; Easy edge-peel packaging; Quick-turn leak-resistant cap; High-quality Puritan polyester tipped swab; Swab is securely attached to cap; Pre-filled with 4ml Neutralizing Buffer; Available in bulk case pack; Shlef life - 18 months; Made in USA
Specifications
Specifications
Sampling Kit
Sterile
Environmental surface sampling with pre-filled neutralizing buffer
Prefilled neutralizing buffer
300 Cs.
103 cm
Plastic
4 mL
4 in.
0.13 oz.
ISO 9001:2008, ISO 13485:2003
Product Suggestions
Product Suggestions