missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puritan™ ESK™ Sampling Kits with Pre-filled Buffered Peptone Water
For environmental surface sampling with buffered peptone water
Supplier: Puritan™ 25-83010PDBPW
Description
For environmental surface sampling; Intended as a nonselective media for the pre-enrichment of Salmonella spp. from food sources; Easy edge-peel packaging; Quick-turn leak-resistant cap; High-quality Puritan polyester tipped swab; Swab is securely attached to cap; Pre-filled with 10ml Buffered Peptone Water; Available in bulk case pack; Shelf life - 18 months; Made in USASpecifications
| Sampling Kit | |
| Sterile | |
| Environmental surface sampling with buffered peptone water and detection of microorganisms | |
| 4mL or 10mL Butterfield's solution, 0.1% peptone and 2% Tween 80 | |
| 50 Cs. | |
| 103 cm |
| Plastic | |
| 10 mL | |
| 4 in. | |
| 0.33 oz. | |
| ISO 9001:2008, ISO 13485:2003 |