Learn More
Sonoco™ PureTemp™ Phase Change Materials
PureTemp™PCMs liquefy or solidify at precise temperatures, from -40° to +37°C. The surrounding temperature falls below the phase temperature, they solidify and release their stored energy. If the temperature rises above the phase temperature, they liquefy and absorb excess energy.
Supplier:  Sonoco™ B27G
 View more versions of this product
Catalog No. 03-600-112
Only null left
Explore available promotions
Description
Description
When the surrounding temperature falls below the phase temperature, PureTemp Phase Change Materials solidify and release their stored energy. If the temperature rises above the phase temperature, they liquefy and absorb excess energy.
  • Biodegradable
  • USDA BioPreferred® designation
  • Extensive range of temperature options
  • The constant temperature during freezing or melting
  • Provides constant temperature control
  • Eliminates thermal shock with fewer excursions
  • Increases payload efficiency
  • Reduces shipping weight and freight costs.
  • Less PureTemp material is required for temperature control when compared to water-based refrigerants
  • Simplifies pack out inventory and process
  • Eliminates bench-time
  • Increases shipping duration
Specifications
Specifications
16/Cs.
1 in.
2.54 cm
20.32 cm
29.2 oz.
0.75 in.
8 x 8 x 1 in.
1°C
Phase Change Material
1°C
8 in.
Rigid Bottle
0.68 kg
0.39 cm
33°F
Product Suggestions
Product Suggestions