missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Tychem QC Disposable Drawstring Hood

Catalog No. 18444011
Encompass
Change view
Click to view available options
Hood; Color: White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18444011 Hood; Color: White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18444011 Supplier Gentex™ Supplier No. PF300003022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pack of 10 disposable Tychem QC drawstring hoods for use with the PureFlo 3000 PAPR.

  • Tychem QC Disposable Drawstring Hood
  • Extra Protection for PureFlo 3000 PAPR

Specifications

Description Tychem QC Disposable Drawstring Hood (10-pack)
Color White
Material Tychem
Product Type Drawstring Hood
Size One Size
For Use With (Application) Extra Protection for PureFlo 3000 PAPR
For Use With (Equipment) PureFlo 3000 PAPR
Model PF3000-03-022
Quantity 10/Pk.
Sterility Non-sterile