missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Standard Faceseal

Catalog No. 18444017
Encompass
Change view
Click to view available options
Face Seal; For use with: PureFlo 3000 PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18444017 Face Seal; For use with: PureFlo 3000 PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18444017 Supplier Gentex™ Supplier No. PF300003054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standard face seal for use with the PureFlo 3000 PAPR.

  • Face Seal for PureFlo 3000 PAPR
  • Black
  • Dartex Material

Specifications

Description Standard faceseal (non welding)
Color Black
Disposable Reusable
Material Dartex Material, Not FR
Product Type Faceseal
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Model PF3000-03-054
Quantity 1 Ea.
Sterility Non-sterile