Learn More
Gentex™ PureFlo 3000 Respirator, Blue Hard Hat
Description
PF3000 Respirator, Blue Hard Hat, Clear Visor, Tychem QC Face Seal No Battery Charger
- PF3000 Respirator
- Blue Hard Hat
- Clear Visor
- Tychem QC Face Seal
- No Battery Charger
Specifications
Specifications
| Assigned Protection Factor (APF) | 25 |
| Cartridge/Filter | PARTICULATE |
| Color | Gray |
| Product Type | PAPR Hard Hat System |
| Size | One Size |
| Intrinsic Safety (IS) Rating | Yes |
| IP Rating | IP54 |