Learn More
Gente™ PureFlo 3000 Respirator with Blue Hard Hat and Clear Visor
Catalog No. 17720122
Change view
Click to view available options
Gente™ Respirator with Blue Hard Hat
Catalog No.
17720122 Gente™ Respirator with Blue Hard Hat
1 options
Catalog No. 17720122 Supplier Gente™ Supplier No. PF3000H2SB03NBC
Only null left
PureFlo 3000 respirator with blue hard hat and clear visor. Standard face seal.
  • PureFlo 3000 respirator with blue hard hat and clear visor.
  • Standard face seal with no battery charger.
Specifications
Assigned Protection Factor (APF) 25
Cartridge/Filter PARTICULATE
Color Gray
Product Type PAPR Hard Hat System
Size One Size
Filter Efficiency (NIOSH) HEPA
Intrinsic Safety (IS) Rating No
IP Rating IP54