missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Power Source Replacement Parts and Accessories DFS Item

Choose from a variety of power sources and associated accessories designed for the PureFlo 3000 PAPR system.

$114.17 - $1,096.10

Specifications

Color Black
Disposable Reusable
For Use With (Equipment) PureFlo 3000 PAPR
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Material Size Price Quantity  
Catalog Number Mfr. No. Product Type Material Size Price Quantity  
18444007 DFS Item
View Documents
Gentex™
PF300003003
Battery Pack Lithium-ion N/A
Only null left
 
18444016 DFS Item
View Documents
Gentex™
PF300003047
Battery Charger Plastic N/A
Only null left
 
18444009 DFS Item
View Documents
Gentex™
PF300003015
Adapter Cable 12 x 7 N/A
Only null left
 
18444030 DFS Item
View Documents
Gentex™
PF300004021
Adapter Cable One Size N/A
Only null left
 
Description

Description

  • Power source replacement parts and accessories for the PureFlo 3000 PAPR system include
    • Rechargeable Lithium Ion battery
    • AC power supply cable North America
    • Battery charger
    • 12V DC Adapter Cable
    • Battery multicharger - 10 station
Specifications

Specifications

Black
Reusable
PureFlo 3000 PAPR
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'