missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentex™ PureFlo 3000 PAPR
NIOSH-approved air-purifying protection systems offer all-in-one respiratory, eye, face, and head protection. Enhanced features for maximum mobility and simplified use make it ideal in pharmaceutical manufacturing and cleanroom environments.
$3175.77 - $3244.05
Specifications
| ANSI Standard | HH: ANSI/ISEA Z89.1:2014 Type 1 Class G ; FACESHIELD - ANSI/ISEA Z87.1:2015 Impact Rated (Z87+) |
|---|---|
| Assigned Protection Factor (APF) | 25 |
| Battery Life | 4 to 5 hours |
| Battery Type | Lithium-ion |
| Cartridge/Filter | PARTICULATE |
Description
- Modular, lightweight PAPR that provides protection in light-to-medium duty industrial environments
- All-in-one protection integrates respiratory, eye, and face protection into one easy-to use unit
- Head-mounted design eliminates the need for hoses and belts, eliminating snag hazards in tight quarters
- Used in a wide variety of applications including healthcare, pharmaceutical manufacturing and cleanroom environments
Specifications
| HH: ANSI/ISEA Z89.1:2014 Type 1 Class G ; FACESHIELD - ANSI/ISEA Z87.1:2015 Impact Rated (Z87+) | |
| 4 to 5 hours | |
| PARTICULATE | |
| PAPR Hard Hat System | |
| PF3000-03-044 | |
| HEPA | |
| 1 Ea. | |
| Yes |
| 25 | |
| Lithium-ion | |
| NIOSH Approved | |
| One Size | |
| High Efficiency Particulate Air (HEPA) Filter | |
| Less than 80 dBA | |
| 2 hr. | |
| Yes |