missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Hood Attachment Blanking Caps

Catalog No. 18444022
Encompass
Change view
Click to view available options
Blanking Caps; For Use With: PureFlo 3000; Includes: Snap Fasteners
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18444022 Blanking Caps; For Use With: PureFlo 3000; Includes: Snap Fasteners
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18444022 Supplier Gentex™ Supplier No. PF300003065

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1 set of hood attachment blanking caps for the PureFlo 3000.

  • Blanking Caps for use with PureFlo 3000 PAPR
  • Includes Snap Fasteners

Specifications

Description Hood Attachment Blanking Caps (set of 2)
Color White
Disposable Reusable
Material Plastic
Product Type Blanking Caps
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Includes Snap Fasteners
Latex Free Yes
Model PF3000-03-065
Quantity 2/Pk.
Sterility Non-sterile
Show More Show Less