missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Faceshield Retaining Clips

Catalog No. 18444010
Encompass
Change view
Click to view available options
Retaining Clips; Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18444010 Retaining Clips; Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18444010 Supplier Gentex™ Supplier No. PF300003016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Set of 2 PureFlo 3000 Faceshield Retaining Clips for left and right sides.

  • Faceshield Retaining Clips
  • To Retain the PF3000 Face Shield

Specifications

Color Black
For Use With (Equipment) PureFlo 3000 PAPR
For Use With (Application) To Retain the PF3000 Face Shield
Material Plastic
Model PF3000-03-016
Mounting Type Clip In
Product Type Clips
Quantity 2/Pk.
Ventilated Not Ventilated