missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo 3000 Air Inlet Adaptor DFS Item

Air inlet adaptors keeps air-purifying protection systems running in peak condition. For use with the PureFlo 3000 PAPR.

$71.45 - $1724.56

Specifications

Color White
Disposable Single-use
Material Plastic
Product Type Air Inlet Adapter
For Use With (Equipment) PureFlo 3000 PAPR
View More Specs

Products 2
Catalog Number Mfr. No. Model Quantity Price Quantity  
Catalog Number Mfr. No. Model Quantity Price Quantity  
18444023 DFS Item
View Documents
Gentex™
PF300003066BP
PF3000-03-066 BP 10/Pk.
Pack of 10 Each for $71.45
Only null left
 
18444024 DFS Item
View Documents
Gentex™
PF300003066MP
PF3000-03-066 MP 250/Pk.
Pack of 250 Each for $1,724.56
Only null left
 
Description

Description

  • For use with PureFlo 3000 PAPR
  • Color: White
Specifications

Specifications

White
Plastic
PureFlo 3000 PAPR
Non-sterile
Single-use
Air Inlet Adapter
Yes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'