missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Amine Rectangular Canted Neck Cell Culture Flask with Vented Cap
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces.
- These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Material | Polystyrene |
| For Use With (Application) | Preparative cell culture, cell-based assays, HTS screening assays |
| Height (English) | 8.66 in. |
| Neck Style | Canted |
| No. per Case | 50 Flasks |
| No. per Pack | 5 Flasks |
| Product Type | Cell Culture Flask |
| Shape | Rectangular |
| Surface Coating | Amine |
| Writing Surface | No |
| Show More |