missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ 96-Well, Amine-Treated, Flat-Bottom Microplate
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
$448.16 - $3285.86
Specifications
| Format | Standard |
|---|---|
| Lid | With Lid |
| Array | 8 x 12 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|
08-772-159
|
Corning™
354717 |
5/Cs |
Case of 5 Each for $448.16
|
|
|||||
|
08-772-169
|
Corning™
356717 |
50/Cs |
Case of 50 Each for $3,285.86
|
|
|||||
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
| Standard | |
| 8 x 12 | |
| Polystyrene | |
| Sterile | |
| Flat Bottom (F-well) | |
| 5/Pk. |
| With Lid | |
| Black, Clear Bottom | |
| 96 | |
| Amine | |
| Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested |