Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 16 to 20 in. wide
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
$4932.91 - $12913.07
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | COC |
| Closure Type | Safe Seal |
| Color | Clear, White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-006-079
|
Keystone Cleanroom Products
6STKRP2048SS |
20 x 48 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-080
|
Keystone Cleanroom Products
6STKRP2046SS |
20 x 46 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-081
|
Keystone Cleanroom Products
6STKRP2044SS |
20 x 44 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-082
|
Keystone Cleanroom Products
6STKRP2040SS |
20 x 40 |
Case of 250 Each for $7,980.15
|
|
|||||
|
17-006-083
|
Keystone Cleanroom Products
6STKRP2036SS |
20 x 36 |
Case of 250 Each for $7,814.22
|
|
|||||
|
17-006-084
|
Keystone Cleanroom Products
6STKRP2032SS |
20 x 32 |
Case of 250 Each for $7,784.04
|
|
|||||
|
17-006-085
|
Keystone Cleanroom Products
6STKRP2030SS |
20 x 30 |
Case of 250 Each for $7,784.04
|
|
|||||
|
17-006-086
|
Keystone Cleanroom Products
6STKRP2026SS |
20 x 26 |
Case of 250 Each for $6,728.07
|
|
|||||
|
17-006-087
|
Keystone Cleanroom Products
6STKRP2024SS |
20 x 24 |
Case of 250 Each for $7,784.04
|
|
|||||
|
17-006-088
|
Keystone Cleanroom Products
6STKRP2020SS |
20 x 20 |
Case of 250 Each for $6,064.30
|
|
|||||
|
17-006-089
|
Keystone Cleanroom Products
6STKRP1870SS |
18 x 70 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-090
|
Keystone Cleanroom Products
6STKRP1854SS |
18 x 54 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-091
|
Keystone Cleanroom Products
6STKRP1848SS |
18 x 48 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-092
|
Keystone Cleanroom Products
6STKRP1840SS |
18 x 40 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-093
|
Keystone Cleanroom Products
6STKRP1836SS |
18 x 36 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-094
|
Keystone Cleanroom Products
6STKRP1832SS |
18 x 32 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-095
|
Keystone Cleanroom Products
6STKRP1830SS |
18 x 30 |
Case of 250 Each for $8,146.10
|
|
|||||
|
17-006-096
|
Keystone Cleanroom Products
6STKRP1828SS |
18 x 28 |
Case of 250 Each for $8,100.83
|
|
|||||
|
17-006-097
|
Keystone Cleanroom Products
6STKRP1824SS |
18 x 24 |
Case of 250 Each for $7,904.73
|
|
|||||
|
17-006-098
|
Keystone Cleanroom Products
6STKRP1820SS |
18 x 20 |
Case of 250 Each for $7,784.04
|
|
|||||
|
17-006-099
|
Keystone Cleanroom Products
6STKRP1648SS |
16 x 48 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-100
|
Keystone Cleanroom Products
6STKRP1644SS |
16 x 44 |
Case of 250 Each for $8,070.66
|
|
|||||
|
17-006-101
|
Keystone Cleanroom Products
6STKRP1638SS |
16 x 38 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17-006-102
|
Keystone Cleanroom Products
6STKRP1636SS |
16 x 36 |
Case of 250 Each for $8,146.10
|
|
|||||
|
17-006-103
|
Keystone Cleanroom Products
6STKRP1635SS |
16 x 35 |
Case of 250 Each for $8,100.83
|
|
|||||
|
17-006-104
|
Keystone Cleanroom Products
6STKRP1632SS |
16 x 32 |
Case of 250 Each for $7,980.15
|
|
|||||
|
17-006-105
|
Keystone Cleanroom Products
6STKRP1630SS |
16 x 30 |
Case of 250 Each for $7,814.22
|
|
|||||
|
17-006-106
|
Keystone Cleanroom Products
6STKRP1628SS |
16 x 28 |
Case of 250 Each for $7,693.52
|
|
|||||
|
17-006-107
|
Keystone Cleanroom Products
6STKRP1624SS |
16 x 24 |
Case of 250 Each for $7,572.85
|
|
|||||
|
17-006-108
|
Keystone Cleanroom Products
6STKRP1620SS |
16 x 20 |
Case of 250 Each for $7,210.80
|
|
|||||
|
17-006-109
|
Keystone Cleanroom Products
6STKRP1616SS |
16 x 16 |
Case of 250 Each for $6,848.74
|
|
|||||
|
17-006-110
|
Keystone Cleanroom Products
6STKRP1612SS |
16 x 12 |
Case of 250 Each for $6,562.13
|
|
|||||
|
17-006-111
|
Keystone Cleanroom Products
6STKRP1604SS |
16 x 4 |
Case of 250 Each for $4,932.91
|
|
|||||
Description
- Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
- Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
Specifications
| Autoclavable | |
| Safe Seal | |
| Single-use | |
| Tyvek | |
| 5.3 mil | |
| 121 to 125 C | |
| Autoclave Bag |
| COC | |
| Clear, White | |
| Steam Sterilization of Pharmaceutical Equipment | |
| Non-sterile | |
| Yes | |
| 5.3 mil |