Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 10 to 15 in. wide
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
$5,731.74 - $9,173.81
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | COC |
| Closure Type | Safe Seal |
| Color | Clear, White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|
17006119
|
Keystone Cleanroom Products
6STKRP1424SS |
14 x 24 |
Case of 250 Each for $7,323.89
|
|
|||||
|
17006120
|
Keystone Cleanroom Products
6STKRP1420SS |
14 x 20 |
Case of 250 Each for $7,248.06
|
|
|||||
|
17006121
|
Keystone Cleanroom Products
6STKRP1418SS |
14 x 18 |
Case of 250 Each for $7,081.27
|
|
|||||
|
17006122
|
Keystone Cleanroom Products
6STKRP1416SS |
14 x 16 |
Case of 250 Each for $7,035.78
|
|
|||||
|
17006123
|
Keystone Cleanroom Products
6STKRP1414SS |
14 x 14 |
Case of 250 Each for $6,884.15
|
|
|||||
|
17006124
|
Keystone Cleanroom Products
6STKRP1248SS |
12 x 48 |
Case of 250 Each for $7,248.06
|
|
|||||
|
17006125
|
Keystone Cleanroom Products
6STKRP1246SS |
12 x 46 |
Case of 250 Each for $7,081.27
|
|
|||||
|
17006126
|
Keystone Cleanroom Products
6STKRP1242SS |
12 x 42 |
Case of 250 Each for $6,959.96
|
|
|||||
|
17006127
|
Keystone Cleanroom Products
6STKRP1240SS |
12 x 40 |
Case of 250 Each for $6,884.15
|
|
|||||
|
17006128
|
Keystone Cleanroom Products
6STKRP1236SS |
12 x 36 |
Case of 250 Each for $6,717.36
|
|
|||||
|
17006129
|
Keystone Cleanroom Products
6STKRP1234SS |
12 x 34 |
Case of 250 Each for $6,505.07
|
|
|||||
|
17006130
|
Keystone Cleanroom Products
6STKRP1232SS |
12 x 32 |
Case of 250 Each for $6,429.24
|
|
|||||
|
17006131
|
Keystone Cleanroom Products
6STKRP1230SS |
12 x 30 |
Case of 250 Each for $6,353.43
|
|
|||||
|
17006132
|
Keystone Cleanroom Products
6STKRP1228SS |
12 x 28 |
Case of 250 Each for $6,262.44
|
|
|||||
|
17006133
|
Keystone Cleanroom Products
6STKRP1224SS |
12 x 24 |
Case of 250 Each for $6,186.63
|
|
|||||
|
17006134
|
Keystone Cleanroom Products
6STKRP1222SS |
12 x 22 |
Case of 250 Each for $5,853.04
|
|
|||||
|
17006135
|
Keystone Cleanroom Products
6STKRP1220SS |
12 x 20 |
Case of 250 Each for $5,853.04
|
|
|||||
|
17006136
|
Keystone Cleanroom Products
6STKRP1218SS |
12 x 18 |
Case of 250 Each for $5,853.04
|
|
|||||
|
17006137
|
Keystone Cleanroom Products
6STKRP1216SS |
12 x 16 |
Case of 250 Each for $5,807.55
|
|
|||||
|
17006138
|
Keystone Cleanroom Products
6STKRP1214SS |
12 x 14 |
Case of 250 Each for $5,777.23
|
|
|||||
|
17006139
|
Keystone Cleanroom Products
6STKRP1212SS |
12 x 12 |
Case of 250 Each for $5,731.74
|
|
|||||
|
17006140
|
Keystone Cleanroom Products
6STKRP1062SS |
10 x 62 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006141
|
Keystone Cleanroom Products
6STKRP1056SS |
10 x 56 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006142
|
Keystone Cleanroom Products
6STKRP1048SS |
10 x 48 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006143
|
Keystone Cleanroom Products
6STKRP1036SS |
10 x 36 |
Case of 250 Each for $6,717.36
|
|
|||||
|
17006144
|
Keystone Cleanroom Products
6STKRP1030SS |
10 x 30 |
Case of 250 Each for $7,414.86
|
|
|||||
|
17006145
|
Keystone Cleanroom Products
6STKRP1024SS |
10 x 24 |
Case of 250 Each for $6,095.65
|
|
|||||
|
17006146
|
Keystone Cleanroom Products
6STKRP1022SS |
10 x 22 |
Case of 250 Each for $5,898.53
|
|
|||||
|
17006147
|
Keystone Cleanroom Products
6STKRP1020SS |
10 x 20 |
Case of 250 Each for $5,807.55
|
|
|||||
|
17006148
|
Keystone Cleanroom Products
6STKRP1018SS |
10 x 18 |
Case of 250 Each for $5,807.55
|
|
|||||
|
17006149
|
Keystone Cleanroom Products
6STKRP1015SS |
10 x 15 |
Case of 250 Each for $5,731.74
|
|
|||||
|
17006112
|
Keystone Cleanroom Products
6STKRP1543SS |
15 x 43 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006113
|
Keystone Cleanroom Products
6STKRP1526SS |
15 x 26 |
Case of 250 Each for $7,323.89
|
|
|||||
|
17006114
|
Keystone Cleanroom Products
6STKRP1448SS |
14 x 48 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006115
|
Keystone Cleanroom Products
6STKRP1440SS |
14 x 40 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006116
|
Keystone Cleanroom Products
6STKRP1438SS |
14 x 38 |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17006117
|
Keystone Cleanroom Products
6STKRP1436SS |
14 x 36 |
Case of 250 Each for $7,657.48
|
|
|||||
|
17006118
|
Keystone Cleanroom Products
6STKRP1426SS |
14 x 26 |
Case of 250 Each for $7,581.67
|
|
|||||
Description
- Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
- Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
Specifications
| Autoclavable | |
| Safe Seal | |
| Single-use | |
| Tyvek | |
| Yes | |
| 5.3 mil | |
| 5.3 mil |
| COC | |
| Clear, White | |
| Steam Sterilization of Pharmaceutical Equipment | |
| Non-sterile | |
| 121 to 125 C | |
| Autoclave Bag |