missing translation for 'onlineSavingsMsg'
Learn More

Biotix Pure uTIP™ Sterile Pipette Tips
GREENER_CHOICE

Catalog No. 12111158
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
250 μL
300 μL
1000 μL
Tip Style:
Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
12111158 300 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12111161 10 μL Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
12111162 10 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12111140 10 μL Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style
12111160 200 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12111159 250 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12111157 1000 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 12111158 Supplier Biotix Supplier No. 63305416
Only null left

Pure uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard are designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Manufactured with Pure resin made of virgin polypropylene.

  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard
  • Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal.
  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin made of virgin polypropylene.
  • Fits all standard pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Hinged Rack Sterile
Tip Style Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Product Type Pipette Tips
Color Code Green
Disposable Single-use
For Use With (Equipment) Universal Fit, Universal Fit Multichannel
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less

This product contains no safety hazards