missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotix Pure uTIP™ Sterile Pipette Tips
Description
- Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard
- Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal.
- Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin made of virgin polypropylene.
- Fits all standard pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Format | Hinged Rack Sterile |
| Tip Style | Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style |
| Product Type | Pipette Tips |
| Color Code | Red |
| Disposable | Single-use |
| Show More |
This product contains no safety hazards