missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotix Pure uTIP™ Bulk Non-sterile Pipette Tips
Description
- Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
- Packaged in re-sealable bags making storage and racking simple.
- Biotix™ uTIP™ Pure resin Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
- Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
- Achieved through patented technologies including FlexFit and Blade
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Re-sealable bag
- Tips must be racked by hand
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Format | Bulk |
| Tip Style | Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point |
| Quantity | 1000 Pk. |
| Product Type | Pipette Tips |
| Color Code | Red |
| Show More |
This product contains no safety hazards