missing translation for 'onlineSavingsMsg'
Learn More

Biotix Pure uTIP™ Bulk Non-sterile Pipette Tips
GREENER_CHOICE

Catalog No. 12111143
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
250 μL
300 μL
1000 μL
1250 μL
Tip Style:
Conductive, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Extended Length, Ergonomic, Finely Tapered, Flextop, Non-beveled Point
Extended Length, Ergonomic, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style
Finely Tapered, Ergonomic, Extended Length, Flextop, Non-beveled Point
Quantity:
1000 Pk.
960 Pk.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Quantity
12111143 1000 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
12111138 10 μL Extended Length, Ergonomic, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style 1000 Pk.
12111148 10 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
12111147 10 μL Finely Tapered, Ergonomic, Extended Length, Flextop, Non-beveled Point 1000 Pk.
12111146 200 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
12111145 250 μL Conductive, Fine Point, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
12111151 300 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point 960 Pk.
12111144 300 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
12111142 1250 μL Extended Length, Ergonomic, Finely Tapered, Flextop, Non-beveled Point 1000 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 12111143 Supplier Biotix Supplier No. 63305431
Only null left

Pure uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk are packaged in re-sealable bags making storage and racking simple. RNase, DNase, Pyrogen, and PCR Inhibitor free.

  • Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
  • Packaged in re-sealable bags making storage and racking simple.
  • Biotix™ uTIP™ Pure resin Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
  • Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
  • Achieved through patented technologies including FlexFit and Blade
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Re-sealable bag
  • Tips must be racked by hand

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bulk, Bagged
Tip Style Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Quantity 1000 Pk.
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Universal Fit, Universal Fit Multichannel
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less

This product contains no safety hazards