missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Pure Solvent Line Kit

Catalog No. 05405486
Change view
Click to view available options
Solvent Line Kit; For: Pure Chromatography Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
05405486 Solvent Line Kit; For: Pure Chromatography Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05405486 Supplier BUCHI Supplier No. 11068215

Pure Chromatography System Solvent Line Kit includes 4 solvent lines and 5 level sensing lines.

  • Includes 4 solvent lines and 5 level sensing lines
  • For use with BUCHI Pure Chromatography Systems

Specifications

Type Solvent Lines
Includes 4 Solvent Lines, 5 Level Sensing Lines
For Use With (Equipment) Pure Chromatography Systems