missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Pure-Fit™ SC Sterile Connectors

Create true sterile connections anywhere – even outside the clean room environment. Saint-Gobain Pure-Fit™ SC secure aseptic connections incorporate Pure-Fit™ SIB™ technology, which provides a seamless transition between tubings and fittings to maintain complete fluid integrity.

Supplier:  Saint-Gobain PFSC250IHM

Encompass

 View more versions of this product

Catalog No. 13417225


Only null left

Description

Description

The silicone valve design at the heart of Pure-Fit™ SC allows for easy assembly and leaves no gaps or residue, eliminating problems associated with other systems. The locking mechanism virtually eliminates the possibility of operator error, and also simplifies validation.

  • Laser etched for complete lot traceability
  • Triple-redundant locking mechanism simplifies validation, reducing chances of operator error
  • Patented Pure-Fit™ SIB™ (smooth inner bore) technology on barb connections ensures a completely smooth fluid path
  • Available in interchangeable 3/4 in. and 1 in. sizes for greater flow volume
  • Available in three interchangeable sizes (1/4 in., 3/8 in., 1/2 in.) for step-down requirements
  • All components made of fully characterized, animal derivative-free and BPA-free materials
  • Easy-to-see laser-etched visual indicators confirm proper engagement
  • Can be sterilized via gamma irradiation or autoclaving
Specifications

Chemical Identifiers

USP Class VI, ISO 10993, EP 3.1.9
32°F
1/4 in. (6.4mm) Hose Barb
104°F
Autoclavable
0°C
35 psi (2.41 bar)
40°C

Specifications

USP Class VI, ISO 10993, EP 3.1.9
PVDF, C-Flex
32°F
1/4 in. (6.4mm) Hose Barb
Red/Clear
104°F
Inner Housing
Autoclavable
4.23 in.
0°C
1.48 in. (3.8cm)
35 psi (2.41 bar)
40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions