Learn More
BUCHI Pure Essential Fraction Collector Door for Funnel Rack
Catalog No. 05405941
Change view
Click to view available options
Collector Door
Catalog No.
05405941 Collector Door
1 options
Catalog No. 05405941 Supplier BUCHI Supplier No. 11075388
This door allows for a closed fraction collector bay while providing a route for collection tubes to exit into bottles or carboys.
  • Enhanced safety when using Pure Essential Funnel Racks
  • Enclose and vent the fraction bay to eliminate exposure risks
  • Place the Pure Essential Chromatography System on a lab bench or in a fume hood
Specifications
Type Collector Door
For Use With (Equipment) Funnel Rack