missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Pure Essential Clamp for Cartridge Stand

Catalog No. 05405942
Change view
Click to view available options
Cartridge Stand Clamp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
05405942 Cartridge Stand Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05405942 Supplier BUCHI Supplier No. 11074604

Use this clamp to hold one or more cartridges ranging from 4 to 300 g.

  • Simple and quick design
  • Securely hold 4to 300 g cartridges
  • Solvent resistant polymer

Specifications

Type Cartridge Holder
For Use With (Equipment) Cartridge Stand