missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Pure Column Holder

Catalog No. 05405476
Change view
Click to view available options
For Use With (Equipment):
10 to 50 mm ID Columns
50 to 70 mm ID Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05405476 10 to 50 mm ID Columns
05405493 50 to 70 mm ID Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05405476 Supplier BUCHI Supplier No. 11066594

Pure Chromatography System Column Holder for use with column diameters ranging from 10 mm to 70 mm.

  • For use with BUCHI Pure Chromatography Systems
  • For column diameters ranging from 10 to 70 mm

Specifications

Type Accessory
For Use With (Application) Holding Columns in Place
For Use With (Equipment) 10 to 50 mm ID Columns
Product Line Pure Chromatography