Learn More
BUCHI Pure Chromatography System
Extremely compact and can be used outside of the fume hood. Choose from flash systems or combination flash/prep HPLC capable of small to large scale purification of complex mixtures. UV detection is standard or choose to add ELS detection to detect all compounds.
Supplier:  BUCHI 11C81500
 View more versions of this product
Catalog No. 10987137
Only null left
Description
Description
  • Pure space. Pure safety. Pure simplicity.
  • Small footprint onboard solvent platform
  • Cartridge/ column holders on front of the system
  • Lowest sample loss for ELSD (30 uL/min)
  • Closed fraction collector bay
  • Leak and pressure sensors for safety
  • Remote control capable from PC, tablet or smartphone
  • All important parts are easily accessable for cleaning
  • Specifications
    Chemical Identifiers
    680 mm
    570 mm
    Specifications
    Separations
    50-60 Hz
    Stainless Steel
    Flash/UV/ELSD
    365 mm
    680 mm
    570 mm
    50 bar (725 psi)
    100/240 V
    40 to 63 μm
    Product Suggestions
    Product Suggestions