missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Pure Cartridge Holders

Catalog No. 05405544
Change view
Click to view available options
For Use With (Equipment):
Cartridges (>300 g)
Cartridges (4 to 330 g)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05405544 Cartridges (>300 g)
05405472 Cartridges (4 to 330 g)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05405544 Supplier BUCHI Supplier No. 11070532

For use with Pure Chromatography Systems

  • For use with BUCHI Pure Chromatography Systems
  • Available in two sizes to accomodate a wide variety of cartridges

Specifications

Type Cartridge Holder
For Use With (Equipment) Cartridges (>300 g)