missing translation for 'onlineSavingsMsg'
Learn More

Pumice Stone, Solstice

Catalog No. 60044856
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044856 500 g Poly Bottle
60044855 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044856 Supplier Solstice Supplier No. 14372500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Granulated

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1332-09-8
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
PubChem CID 76973482
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
SMILES O=[Si]=O.O=[Al]O[Al]=O

Specifications

Physical Form Granulated
Quantity 500 g
Color Gray
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Pumice Stone