missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pumice Stone, Honeywell Fluka™
Granulated
Supplier: Honeywell-Fluka 143721KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| 1 kg | |
| Granulated | |
| Poly Bottle | |
| Al2O5Si | |
| NONH for all modes of transport | |
| O=[Si]=O.O=[Al]O[Al]=O | |
| 162.04 |
| Gray | |
| Pumice Stone | |
| 1332-09-8 | |
| MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733 | |
| HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
| oxo[(oxoalumanyl)oxy]alumane; silanedione | |
| 76973482 |
Chemical Identifiers
| 1332-09-8 | |
| 162.04 | |
| HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
| oxo[(oxoalumanyl)oxy]alumane; silanedione |
| Al2O5Si | |
| MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733 | |
| 76973482 | |
| O=[Si]=O.O=[Al]O[Al]=O |