missing translation for 'onlineSavingsMsg'
Learn More

Pulegone, 92%, pract.

Catalog No. AC418501000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418501000 100 g Glass bottle
AC418505000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418501000 Supplier Thermo Scientific Chemicals Supplier No. 418501000

Pulegone, CAS # 89-82-7, is a compound found in the oils of certain plant species including peppermint, catnip, and pennyroyal. | CAS: 89-82-7 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations

Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation

Chemical Identifiers

CAS 89-82-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00063000
InChI Key NZGWDASTMWDZIW-QMMMGPOBSA-N
Synonym --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
PubChem CID 638012
ChEBI CHEBI:81226
IUPAC Name (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
SMILES CC1CCC(=C(C)C)C(=O)C1

Specifications

Color Colorless to Yellow
Density 0.9310g/mL
Boiling Point 97°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.488
Quantity 100 g
Beilstein 07, 81
Specific Gravity 0.931
Merck Index 15, 8046
Specific Rotation Condition +21° (20°C neat)
Specific Rotation + 21
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,chloroform,diethyl ether
Formula Weight 152.24
Percent Purity 92%
Grade Practical
Physical Form Liquid
Chemical Name or Material Pulegone
Show More Show Less
EINECSNumber 201-943-2

RUO – Research Use Only