missing translation for 'onlineSavingsMsg'
Learn More

Pulegone, 92%, pract.

Pulegone, CAS # 89-82-7, is a compound found in the oils of certain plant species including peppermint, catnip, and pennyroyal. | CAS: 89-82-7 | C10H16O | 152.24 g/mol

$854.45 - $854.45

Chemical Identifiers

CAS 89-82-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00063000
InChI Key NZGWDASTMWDZIW-QMMMGPOBSA-N
Synonym --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
PubChem CID 638012
ChEBI CHEBI:81226
IUPAC Name (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
SMILES CC1CCC(=C(C)C)C(=O)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC418505000
SDS View Documents
Thermo Scientific Chemicals
418505000
500 g Glass bottle
Each for $854.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations

Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation

Specifications

Chemical Identifiers

89-82-7
152.24
NZGWDASTMWDZIW-QMMMGPOBSA-N
638012
(5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
C10H16O
MFCD00063000
--pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
CHEBI:81226
CC1CCC(=C(C)C)C(=O)C1

Specifications

89-82-7
0.9310g/mL
Authentic
Glass bottle
1.4850 to 1.488
500 g
0.931
+21° (20°C neat)
--pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
NZGWDASTMWDZIW-QMMMGPOBSA-N
(5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
638012
152.24
Practical
Pulegone
Colorless to Yellow
97°C (12.0 mmHg)
90% min. (GC)
C10H16O
MFCD00063000
07, 81
15, 8046
+ 21
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,chloroform,diethyl ether
CC1CCC(=C(C)C)C(=O)C1
152.24
CHEBI:81226
92%
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-943-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only