missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUFA No.2, Animal Source, Analytical Standard, MilliporeSigma™ Supelco™
Description
For qualitative identification only. Product is extracted from natural sources. The fatty acid composition varies from lot-to-lot.
Specifications
Specifications
| Physical Form | Neat |
| Synonym | Polyunsaturated Fatty Acid Mix No. 2 |
| Quantity | 100 mg |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Methyl myristate; Methyl palmitate; Methyl palmitoleate; Methyl stearate; Methyl oleate; cis-11-Octadecenoic methyl ester; Methyl linoleate; Methyl linolenate; Methyl gamma-linolenate; Methyl cis-11-eicosenoate; Methyl arachidonate; Methyl all-cis-5,8,11,14,17-eicosapentaenoate; cis-7,10,13,16-Docosatetraenoic acid methyl ester; cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester |