missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PTFE/Silicone Discs

Catalog No. PI12722
Change view
Click to view available options
Diameter (Metric) Outer:
8 mm
12 mm
13 mm
16 mm
18 mm
22 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
PI12722 22 mm
PI12708 8 mm
PI12712 12 mm
PI12713 13 mm
PI12716 16 mm
PI12718 18 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PI12722 Supplier Thermo Scientific™ Supplier No. TS12722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combine the inertness of a PTFE coating with the resealability of silicone Thermo Scientific™ PTFE/Silicone Discs.

Ensure air-tight, inert seals with autoclavable Thermo Scientific™ PTFE/Silicone Discs. They are specifically designed to combine the resealability of silicone with the inertness of PTFE. These discs reseal instantly puncture after puncture, and the PTFE coating is structurally bonded so that no cement is leached or baked out of your sample after needle penetration. Many sizes are available to fit Thermo Scientific™ Reacti-Vial™ Small Reaction Vials, Thermo Scientific™ Screw Cap Septum Vials, and Thermo Scientific™ Hypo-ViaI™ Sample Storage Vials.

Additional Product Information

Each disc compresses to maintain a tight seal, forcing the PTFE coating to conform to the sealing surface.

Standard syringe and CC needles penetrate an entire disc with ease.

The PTFE layer is a full 10 mils thick to ensure protection

Specifications

Description Diameter: 22mm
Diameter (Metric) Outer 22 mm
Material PTFE, Silicone
Quantity 72 Pk.
Thickness (Metric) 10 mil
For Use With (Equipment) 25 and 40 mL Screw Cap Septum Vial, 240 mL (8 oz.) Screw Cap Bottle, 10 mL Reacti-Vial
Autoclavable Autoclavable
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Disc

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.