missing translation for 'onlineSavingsMsg'
Learn More

PTFE sheet, 3.18mm (0.125in) thick

Catalog No. AA45172HB
Change view
Click to view available options
Quantity:
300 x 300 mm
Packaging:
Shrink-wrapped
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA45172HB 300 x 300 mm Shrink-wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45172HB Supplier Thermo Scientific Chemicals Supplier No. 045172HB
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-84-0 | (C2F4)n | NaN g/mol

PTFE sheet is used for the manufacture of high-temperature seals, insulators and bearings used in semiconductor, aerospace and chemical processing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Packaging Shrink-wrapped
Quantity 300 x 300 mm
Solubility Information Insoluble in water.
Chemical Name or Material PTFE sheet
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only