Learn More
Heidolph™ PTFE Seal for Hei-VAP Industrial Rotary Evaporator
Catalog No. 13900961
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13900961 Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13900961 Supplier Heidolph™ Supplier No. 036290033
PTFE seal seals the joint between evaporating flask and vapor tube.
Included with Hei-VAP Industrial Rotary Evaporator
Specifications
Type Seal
For Use With (Equipment) Hei-VAP Industrial Rotary Evaporator