missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PTFE/Rubber Laminated Discs

Catalog No. PI12422
Change view
Click to view available options
Closure Size:
12 mm
18 mm
22 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
PI12422 22 mm
PI12412 12 mm
PI12418 18 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI12422 Supplier Thermo Scientific™ Supplier No. TS12422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For a highly inert and unreactive seal use Thermo Scientific™ PTFE/Rubber Laminated Discs.

For a highly inert and unreactive seal use Thermo Scientific™ PTFE/Rubber Laminated Discs. Constructed of white pharmaceutical rubber with 5 mils of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert nonreactive seal is desired. Discs are autoclavable with no loss of integrity after heating above 100 °C for five hours. Several sizes are available to fit Thermo Scientific™ Reacti-Vial™ Small Reaction Vials and Thermo Scientific™ Screw Cap Septum Vials.

Additional Product Information

Disc thickness is approximately 60 mils

Discs are rigid and more difficult to puncture than Thermo Scientific™ PTFE/Silicone Discs

Specifications

Description 22mm
Material PTFE, Rubber
Quantity 72 Pk.
Thickness (Metric) 60 mil
Closure Size 22 mm
For Use With (Equipment) Small Reaction Vial
Autoclavable Autoclavable
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Disc

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.