Learn More
Thermo Scientific™ PTFE One-Piece HPLC Column Connector
Catalog No. 03251822
Change view
Click to view available options
Material:
PEEK
Catalog No. Material
03251822 PEEK
1 options
Catalog No. 03251822 Supplier Thermo Scientific™ Supplier No. 60170370
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ensure leak-free column connections and optimize liquid chromatography performance with the inert and biocompatible Thermo Scientific™ PTFE One-Piece HPLC Column Connector.
Minimize HPLC system dead volume and ensure optimal separation efficiencies with the Thermo Scientific™ PTFE One-Piece HPLC Column Connector. Ideal for coupling analytical and guard columns with 10–32 threads, it provides fingertight, leak-free connections. The material is inert and biocompatible for a wide range of applications.
Specifications
Type Coupler
Description One-piece column coupler
Material PEEK
For Use With (Equipment) Analytical and Guard Columns with 10 to 32 Threads