missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PTFE-Lined Closed Caps
GREENER_CHOICE

Catalog No. p-4521716
Change view
Click to view available options
Neck Finish:
89-400
Color:
White
Product Type:
Closure with Bonded Liner
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Color Product Type
05719601 89-400 White Closure with Bonded Liner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05719601 Supplier Thermo Scientific™ Supplier No. B1678900BL
Only null left

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

Specifications

Certification Parameters None
Color White
Description Closed top cap
Material Polypropylene
No. per Case 24
Level Unprocessed
Liner Material PTFE
Neck Finish 89-400
Certifications/Compliance Manufactured under an ISO certified quality management system.
Shape Round
Product Type Closure with Bonded Liner
Show More Show Less