Learn More
Celltreat Scientific Products PTFE Syringe Filter, 0.22m, 13mm, Sterile
Supplier:  Celltreat Scientific Products 229756
Encompass
CELLTREAT Scientific Products 0.22m PTFE Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. PTFE (Polytetrafluoroethylene). Hydrophobic membrane has great temperature resistance. Degassing/clarifying aqueous samples. Strong acid solvent filtration. Alkali solvent filtration. Membrane diameter 13mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister packs. 75/case.
Catalog No. 50202071
Only null left